Learn More
Biomatik Corporation Horse Leptin (Lep) Elisa Kit
Supplier:  Biomatik Corporation EKC32359
Encompass_Preferred
Horse Leptin (Lep) Elisa Kit for detecting Leptin in serum, plasma, tissue homogenates. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q9TU09; LEP. Target Synonyms: LEP; OB; Leptin; Obesity factor; Fragment. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3744