Learn More
Biomatik Corporation Human 17-Hydroxyprogesterone, 17-Ohp Elisa Kit
Supplier:  Biomatik Corporation EKC32387
Encompass_Preferred
Human 17-Hydroxyprogesterone, 17-Ohp Elisa Kit for detecting 17-Hydroxyprogesterone, 17-Ohp in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.3125ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3752