Learn More
Biomatik Corporation Human A Disintegrin And Metalloprotease 10, Adam10 Elisa Kit
Supplier:  Biomatik Corporation EKC32425
Encompass_Preferred
Human A Disintegrin And Metalloprotease 10, Adam10 Elisa Kit for detecting Adam Metallopeptidase Domain 10 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O14672; ADAM10. Target Synonyms: ADAM10; KUZ; MADM; Disintegrin and metalloproteinase domain-containing protein 10; ADAM 10; CDw156; Kuzbanian protein homolog; Mammalian disintegrin-metalloprotease; CD antigen CD156c. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3760