Learn More
Biomatik Corporation Human Acetyl-Coa Carboxylase 1 (Acaca) Elisa Kit
Supplier:  Biomatik Corporation EKC32457
Encompass_Preferred
Human Acetyl-Coa Carboxylase 1 (Acaca) Elisa Kit for detecting Acetyl-Coenzyme A Carboxylase Alpha in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 9.38-600pg/ml. Sensitivity: 2.34pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13085; ACACA. Target Synonyms: ACAC; ACACA; ACACA_HUMAN; ACACB; ACC alpha; ACC; ACC beta; ACC-alpha; ACC1; ACC2; ACCA; ACCB; Acetyl CoA carboxylase 1; Acetyl CoA carboxylase 2; Acetyl CoA carboxylase alpha; Acetyl CoA carboxylase beta; Acetyl Coenzyme A carboxylase alpha; Acetyl Coenzyme A carboxylase beta; Biotin carboxylase; COA1; COA2; HACC275; OTTHUMP00000164069; OTTHUMP00000164070; OTTHUMP00000164076; OTTHUMP00000240532. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3766