Learn More
Biomatik Corporation Human Activity-Regulated Cytoskeleton-Associated Protein (Arc) Elisa Kit
Supplier:  Biomatik Corporation EKC32477
Encompass_Preferred
Human Activity-Regulated Cytoskeleton-Associated Protein (Arc) Elisa Kit for detecting Activity-Regulated Cytoskeleton-Associated Protein in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q7LC44; ARC. Target Synonyms: activity regulated cytoskeleton associated protein; activity regulated gene 3.1 protein homolog; Activity-regulated cytoskeleton-associated protein; Activity-regulated gene 3.1 protein homolog; Arc; ARC/ARG3.1; ARC_HUMAN; Arg 3.1; Arg3.1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3768