Learn More
Biomatik Corporation Human Adropin (Enho) Elisa Kit
Supplier:  Biomatik Corporation EKC32507
Encompass_Preferred
Human Adropin (Enho) Elisa Kit for detecting Energy Homeostasis Associated in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6UWT2; ENHO. Target Synonyms: Adropin; C9orf165; Energy homeostasis associated protein; Energy homeostasis-associated protein; ENHO; ENHO_HUMAN; PRO830; UNQ470. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3779