Learn More
Biomatik Corporation Human Alanine Aminotransferase, Alt/Gpt Elisa Kit
Supplier:  Biomatik Corporation EKC32516
Encompass_Preferred
Human Alanine Aminotransferase, Alt/Gpt Elisa Kit for detecting Glutamic-Pyruvate Transaminase (Alanine Aminotransferase) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000uIU/ml. Sensitivity: 3.9uIu/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P24298; GPT. Target Synonyms: AAT1; Alanine aminotransferase 1; Alanine aminotransferase; ALAT1_HUMAN; ALT1; Glutamate pyruvate transaminase 1; Glutamic alanine transaminase 1; Glutamic pyruvate transaminase (alanine aminotransferase); Glutamic pyruvic transaminase 1; Glutamic--alanine transaminase 1; Glutamic--pyruvic transaminase 1; GPT 1; gpt; GPT1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3781