Learn More
Biomatik Corporation Human Anthrax Toxin Receptor 2 (Antxr2) Elisa Kit
Supplier:  Biomatik Corporation EKC32612
Encompass_Preferred
Human Anthrax Toxin Receptor 2 (Antxr2) Elisa Kit for detecting Anthrax Toxin Receptor 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P58335; ANTXR2. Target Synonyms: ANTXR2; CMG2; Anthrax toxin receptor 2; Capillary morphogenesis gene 2 protein; CMG-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3798