Learn More
Biomatik Corporation Human Anti-Muellerian Hormone Type-2 Receptor (Amhr2) Elisa Kit
Supplier:  Biomatik Corporation EKC32692
Encompass_Preferred
Human Anti-Muellerian Hormone Type-2 Receptor (Amhr2) Elisa Kit for detecting Anti-Mullerian Hormone Receptor, Type Ii in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.32-12ng/ml. Sensitivity: 8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q16671; AMHR2. Target Synonyms: AMH type II receptor; AMHR; AMHR2; AMHR2_HUMAN; Anti Mullerian hormone receptor; type II; Anti-Muellerian hormone type II receptor; Anti-Muellerian hormone type-2 receptor; MGC141312; MIS type II receptor; MISR2; MISRII; MRII; Muellerian hormone type 2 receptor; Muellerian hormone type II receptor; Muellerian inhibiting substance type II receptor; Mullerian hormone receptor type II; Mullerian inhibiting substance type II receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3801