Learn More
Biomatik Corporation Human Apelin 12, Ap12 Elisa Kit
Supplier:  Biomatik Corporation EKC32749
Encompass_Preferred
Human Apelin 12, Ap12 Elisa Kit for detecting Apelin 12, Ap12 in serum, plasma, saliva, urine, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3803