Learn More
Biomatik Corporation Human Apelin 13, Ap13 Elisa Kit
Supplier:  Biomatik Corporation EKC32750
Encompass_Preferred
Human Apelin 13, Ap13 Elisa Kit for detecting Apelin 13, Ap13 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3804