Learn More
Biomatik Corporation Human Apoptosis-Associated Speck-Like Protein Containing A Card (Pycard) Elisa Kit
Supplier:  Biomatik Corporation EKC32776
Encompass_Preferred
Human Apoptosis-Associated Speck-Like Protein Containing A Card (Pycard) Elisa Kit for detecting Pyd And Card Domain Containing in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9ULZ3; PYCARD. Target Synonyms: Apoptosis associated speck like protein containing a CARD; Apoptosis-associated speck-like protein containing a CARD; ASC; ASC_HUMAN; CARD 5; CARD5; Caspase recruitment domain containing protein 5; Caspase recruitment domain protein 5; Caspase recruitment domain-containing protein 5; hASC; MGC10332; PYCARD; PYD and CARD domain containing; PYD and CARD domain containing protein; PYD and CARD domain-containing protein; Target of methylation induced silencing 1; Target of methylation-induced silencing 1; TMS 1; TMS; TMS1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3808