Learn More
Biomatik Corporation Human Aquaporin 1, Aqp-1 Elisa Kit
Supplier:  Biomatik Corporation EKC32777
Encompass_Preferred
Human Aquaporin 1, Aqp-1 Elisa Kit for detecting Aquaporin 1 (Colton Blood Group) in serum, plasma, cell culture supernates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P29972; AQP1. Target Synonyms: AQP1; CHIP28; Aquaporin-1; AQP-1; Aquaporin-CHIP; Urine water channel; Water channel protein for red blood cells and kidney proximal tubule. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3809