Learn More
Biomatik Corporation Human Arginase 2 (Arg2) Elisa Kit
Supplier:  Biomatik Corporation EKC32788
Encompass_Preferred
Human Arginase 2 (Arg2) Elisa Kit for detecting Arginase, Type Ii in serum, plasma, cell culture supernates, urine, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 0.47-30mU/ml. Sensitivity: 0.12mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P78540; ARG2. Target Synonyms: ARG2; ARGI2_HUMAN; Arginase II mitochondrial; Arginase type II; Arginase-2; arginase-2; mitochondrial; Kidney arginase; Kidney type arginase; Kidney-type arginase; L arginine amidinohydrolase; L arginine ureahydrolase; mitochondrial; Non hepatic arginase; Non-hepatic arginase; Nonhepatic arginase; Type II arginase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3810