Learn More
Biomatik Corporation Human Asialoglyco Protein Receptor, Asgpr Elisa Kit
Supplier:  Biomatik Corporation EKC32799
Encompass_Preferred
Human Asialoglyco Protein Receptor, Asgpr Elisa Kit for detecting Asialoglyco Protein Receptor, Asgpr in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2000ng/ml. Sensitivity: 7.8ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07306; ASGR1. Target Synonyms: ASGP-R 1; ASGPR 1; ASGPR; ASGPR1; Asgr1; ASGR1_HUMAN; Asialoglycoprotein receptor 1; C type lectin domain family 4 member H1; C-type lectin domain family 4 member H1; CLEC4H1; Hepatic lectin H1; HL-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3811