Learn More
Biomatik Corporation Human B1 Bradykinin Receptor (Bdkrb1) Elisa Kit
Supplier:  Biomatik Corporation EKC32822
Encompass_Preferred
Human B1 Bradykinin Receptor (Bdkrb1) Elisa Kit for detecting Bradykinin Receptor B1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P46663; BDKRB1. Target Synonyms: BDKRB1; BRADYB1; B1 bradykinin receptor; B1R; BK-1 receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3815