Learn More
Biomatik Corporation Human B7-H4 Elisa Kit
Supplier:  Biomatik Corporation EKC32823
Encompass_Preferred
Human B7-H4 Elisa Kit for detecting V-Set Domain Containing T Cell Activation Inhibitor 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500ng/ml. Sensitivity: 1.95ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q7Z7D3; VTCN1. Target Synonyms: B7 family member; H4; B7 H4; B7 homolog 4; B7 superfamily member 1; B7 superfamily; member 1; B7-H4; B7h.5; B7h4; B7S1; B7x; BC032925; Immune costimulatory protein B7-H4; Immune costimulatory protein B7H4; MGC41287; PRO1291; Protein B7S1; RP11 229A19.4; T cell costimulatory molecule B7x; T-cell costimulatory molecule B7x; V set domain-containing T cell activation inhibitor 1; V-set domain-containing T-cell activation inhibitor 1; VCTN1; Vtcn1; VTCN1_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3816