Learn More
Biomatik Corporation Human Bactericidal/Permeabilityincreasingprotein, Bpi Elisa Kit
Supplier:  Biomatik Corporation EKC32827
Encompass_Preferred
Human Bactericidal/Permeabilityincreasingprotein, Bpi Elisa Kit for detecting Bactericidal/Permeability-Increasing Protein in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P17213; BPI. Target Synonyms: Bactericidal permeability-increasing protein; Bactericidal/permeability increasing protein; BPI; BPI fold containing family D; member 1; BPI_HUMAN; BPIFD1; CAP 57; CAP57; rBPI; Recombinant BPI holoprotein; rBPI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3817