Learn More
Biomatik Corporation Human Baculoviral Iap Repeat-Containing Protein 2 (Birc2) Elisa Kit
Supplier:  Biomatik Corporation EKC32828
Encompass_Preferred
Human Baculoviral Iap Repeat-Containing Protein 2 (Birc2) Elisa Kit for detecting Baculoviral Iap Repeat-Containing 2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13490; BIRC2. Target Synonyms: API 1; API1; Apoptosis inhibitor 1; Baculoviral IAP repeat containing 2; Baculoviral IAP repeat containing protein 2; Baculoviral IAP repeat-containing protein 2; BIRC 2; BIRC2; C-IAP1; Cellular inhibitor of apoptosis 1; cellular inhibitor of apoptosis protein 1; cIAP 1; cIAP1; HIAP 2; HIAP-2; HIAP2; IAP 2; IAP homolog B; IAP-2; IAP2; Inhibitor of apoptosis protein 2; MIHB; NFR2 TRAF signalling complex protein; RING finger protein 48; RNF 48; RNF48; TNFR2 TRAF signaling complex protein 2; TNFR2-TRAF-signaling complex protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3818