Learn More
Biomatik Corporation Human Bcl-2-Like Protein 11 (Bcl2L11) Elisa Kit
Supplier:  Biomatik Corporation EKC32842
Encompass_Preferred
Human Bcl-2-Like Protein 11 (Bcl2L11) Elisa Kit for detecting Bcl2-Like 11 (Apoptosis Facilitator) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.44-1500pg/ml. Sensitivity: 5.86pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O43521; BCL2L11. Target Synonyms: BCL2 like 11; B2L11_HUMAN; BAM; Bcl 2 interacting protein Bim; Bcl 2 related ovarian death agonist; Bcl-2-like protein 11; BCL2 interacting mediator of cell death; BCL2 like 11 (apoptosis facilitator); BCL2 like protein 11; Bcl2-interacting mediator of cell death; Bcl2-L-11; Bcl2l11; BIM alpha6; BIM; BIM beta6; BIM beta7; BimEL; BimL; BOD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3822