Learn More
Biomatik Corporation Human Apolipoprotein H (Apoh) Elisa Kit
Supplier:  Biomatik Corporation EKC32849
Encompass_Preferred
Human Apolipoprotein H (Apoh) Elisa Kit for detecting Apolipoprotein H(Apoh) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000ng/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02749; APOH. Target Synonyms: Activated protein C binding protein; Activated protein C-binding protein; Anticardiolipin cofactor; APC inhibitor; Apo-H; APOH; APOH_HUMAN; apolipoprotein H (beta-2-glycoprotein I); Apolipoprotein H; B2G1; B2GP1; B2GPI; Beta 2 glycoprotein I; Beta 2 glycoprotein I precursor; Beta (2)GPI; Beta-2-glycoprotein 1; Beta-2-glycoprotein I; BG; Glycoprotein 1; beta-2; Glycoprotein I; beta-2; OTTMUSP00000003033. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3824