Learn More
Biomatik Corporation Human Bone Morphogenetic Protein Receptor 1A, Bmpr-1A Elisa Kit
Supplier:  Biomatik Corporation EKC32899
Encompass_Preferred
Human Bone Morphogenetic Protein Receptor 1A, Bmpr-1A Elisa Kit for detecting Bone Morphogenetic Protein Receptor, Type Ia in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P36894; BMPR1A. Target Synonyms: 10q23del; Activin A receptor type II like kinase 3; Activin receptor like kinase 3; Activin receptor-like kinase 3; ACVRLK 3; ACVRLK3; ALK 3; ALK-3; ALK3; BMP type-1A receptor; BMPR 1A; Bmpr; BMPR-1A; Bmpr1a; BMR1A_HUMAN; Bone morphogenetic protein receptor type IA; Bone morphogenetic protein receptor type IA precursor; Bone morphogenetic protein receptor type-1A; BR 1a; BR1a; CD 292; CD292; CD292 antigen; EC 2.7.11.30; Serine threonine protein kinase receptor R5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3830