Learn More
Biomatik Corporation Human Bromodomain Testis-Specific Protein (Brdt) Elisa Kit
Supplier:  Biomatik Corporation EKC32917
Encompass_Preferred
Human Bromodomain Testis-Specific Protein (Brdt) Elisa Kit for detecting Bromodomain, Testis-Specific in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q58F21; BRDT. Target Synonyms: BRDT; Bromodomain testis-specific protein; Cancer/testis antigen 9; CT9; RING3-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3831