Learn More
Biomatik Corporation Human Calbindin (Calb1) Elisa Kit
Supplier:  Biomatik Corporation EKC32940
Encompass_Preferred
Human Calbindin (Calb1) Elisa Kit for detecting Calbindin 1, 28Kda in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05937; CALB1. Target Synonyms: avian-type; CAB27; CALB 1; CALB; CALB1; CALB1_HUMAN; Calbindin 1 28kDa; Calbindin; Calbindin D28; D 28K; D-28K; D28K; OTTHUMP00000166027; OTTHUMP00000225441; RTVL H protein; Vitamin D dependent calcium binding protein; Vitamin D dependent calcium binding protein avian type; Vitamin D-dependent calcium-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3834