Learn More
Biomatik Corporation Human Calcitonin, Ct Elisa Kit
Supplier:  Biomatik Corporation EKC32942
Encompass_Preferred
Human Calcitonin, Ct Elisa Kit for detecting Calcitonin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 4.688-300pg/ml. Sensitivity: 1.172pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01258; CALCA. Target Synonyms: CALCA; CALC1Calcitonin [Cleaved into: Calcitonin; Katacalcin; Calcitonin carboxyl-terminal peptide; CCP; PDN-21)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3835