Learn More
Biomatik Corporation Human Carcinoembryonic Antigen-Related Cell Adhesion Molecule 6 (Ceacam6) Elisa Kit
Supplier:  Biomatik Corporation EKC33006
Encompass_Preferred
Human Carcinoembryonic Antigen-Related Cell Adhesion Molecule 6 (Ceacam6) Elisa Kit for detecting Carcinoembryonic Antigen-Related Cell Adhesion Molecule 6 (Non-Specific Cross Reacting Antigen) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.83-20ng/ml. Sensitivity: 0.33ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P40199; CEACAM6. Target Synonyms: Carcinoembryonic antigen related cell adhesion molecule 6; Carcinoembryonic antigen related cell adhesion molecule 6 (non specific cross reacting antigen); Carcinoembryonic antigen-related cell adhesion molecule 6; CD 66c; CD66c; CD66c antigen; CEA LIKE PROTEIN; CEACAM 6; CEACAM6; CEAL; CEAM6_HUMAN; MGC93832; NCA; Non specific cross reacting antigen; Non-specific crossreacting antigen; Normal cross reacting antigen; Normal cross-reacting antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3845