Learn More
Biomatik Corporation Human Casein Alpha S1 (Csn1S1) Elisa Kit
Supplier:  Biomatik Corporation EKC33025
Encompass_Preferred
Human Casein Alpha S1 (Csn1S1) Elisa Kit for detecting Casein Alpha S1 in breast milk. Species Reactivity: Human (Homo sapiens). Detection Range: 37.5-600ug/ml. Sensitivity: 18.75ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P47710; CSN1S1. Target Synonyms: Alpha casein; alpha-S1-casein; CASA; CASA1_HUMAN; Casein alpha s1; casein; alpha; casein; alpha-S1; included; Casoxin-D; CSN1; CSN1S1; Csna; MGC149368. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3850