Learn More
Biomatik Corporation Human Cd160 Antigen (Cd160) Elisa Kit
Supplier:  Biomatik Corporation EKC33067
Encompass_Preferred
Human Cd160 Antigen (Cd160) Elisa Kit for detecting Cd160 Molecule in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O95971; CD160. Target Synonyms: BY55; BY55_HUMAN; CD160; CD160 antigen [Precursor]; CD160 antigen; CD160 delta Ig; CD160 molecule; CD160 transmembrane isoform; FLJ46513; Natural killer cell receptor BY55; Natural killer cell receptor; immunoglobulin superfamily member; NK1; NK28. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3857