Learn More
Biomatik Corporation Human Cd2-Associated Protein (Cd2Ap) Elisa Kit
Supplier:  Biomatik Corporation EKC33074
Encompass_Preferred
Human Cd2-Associated Protein (Cd2Ap) Elisa Kit for detecting Cd2-Associated Protein in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 18.75-1200pg/ml. Sensitivity: 4.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Y5K6; CD2AP. Target Synonyms: Adapter protein CMS; AL024079; C78928; Cas ligand with multiple SH3 domains; CD2 associated protein; CD2-associated protein; CD2AP; CD2AP_HUMAN; CMS; Mesenchyme to epithelium transition protein with SH3 domains 1; METS 1; Mets1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3859