Learn More
Biomatik Corporation Human Chemokine (C-X-C Motif) Ligand 17 (Cxcl17) Elisa Kit
Supplier:  Biomatik Corporation EKC33109
Encompass_Preferred
Human Chemokine (C-X-C Motif) Ligand 17 (Cxcl17) Elisa Kit for detecting Chemokine (C-X-C Motif) Ligand 17 (Cxcl17) in serum, plasma, cell culture supernates, urine, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6UXB2; CXCL17. Target Synonyms: CXCL17; VCC1; UNQ473/PRO842C-X-C motif chemokine 17; 6-Cys CXCL17; Dendritic cell and monocyte chemokine-like protein; DMC; VEGF coregulated chemokine 1) [Cleaved into: 4-Cys CXCL17]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3862