Learn More
Biomatik Corporation Human cluster Of differentiation 82 (CD82) ELISA Kit
Supplier:  Biomatik Corporation EKC33167
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P27701. Target Synonyms: 4F9, C33, GR15, IA4, KAI1, R2, SAR2, ST6, TSPAN27, C33 antigen|CD82 antigen|R2 leukocyte antigen|inducible membrane protein R2|kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 l. Restriction: For Research Use Only.
Catalog No. 50-229-3871