Learn More
Biomatik Corporation Human Cofilin-1 (Cfl1) Elisa Kit
Supplier:  Biomatik Corporation EKC33187
Encompass_Preferred
Human Cofilin-1 (Cfl1) Elisa Kit for detecting Cofilin 1 (Non-Muscle) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P23528; CFL1. Target Synonyms: 18 kDa phosphoprotein; CFL 1; CFL; CFL1; COF1_HUMAN; Cofilin 1; Cofilin 1 non muscle; Cofilin; Cofilin non muscle isoform; Cofilin-1; epididymis secretory protein Li 15; HEL-S-15; non-muscle isoform; p18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3876