Learn More
Biomatik Corporation Human Collagen Alpha-2 (I) Chain (Col1A2) Elisa Kit
Supplier:  Biomatik Corporation EKC33207
Encompass_Preferred
Human Collagen Alpha-2 (I) Chain (Col1A2) Elisa Kit for detecting Collagen, Type I, Alpha 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P08123; COL1A2. Target Synonyms: Alpha 2 collagen type I; Alpha 2 type I collagen; Alpha 2 type I procollagen; Alpha 2 (I) collagen; Alpha 2 (I) procollagen; Alpha-2 type I collagen; CO1A2_HUMAN; COL1A2; Collagen alpha 2 (I) chain; Collagen alpha-2 (I) chain; Collagen I alpha 2 polypeptide; Collagen of skin tendon and bone alpha 2 chain; Collagen type I alpha 2; OI4; Osteogenesis imperfecta type IV; Type I procollagen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3880