Learn More
Biomatik Corporation Human Collectin-12 (Colec12) Elisa Kit
Supplier:  Biomatik Corporation EKC33234
Encompass_Preferred
Human Collectin-12 (Colec12) Elisa Kit for detecting Collectin Sub-Family Member 12 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q5KU26; COLEC12. Target Synonyms: CL P1; CL-P1; CLP1; COL12_HUMAN; COLEC 12; colec12; Collectin placenta 1; Collectin placenta protein 1; Collectin sub family member 12; Collectin-12; hCL P1; hCL-P1; NSR2; Nurse cell scavenger receptor 2; SCARA4; Scavenger receptor class A member 4; Scavenger receptor with C type lectin; Scavenger receptor with C-type lectin; SRCL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3883