Learn More
Biomatik Corporation Human Complement C1Q Tumor Necrosis Factor-Related Protein 1 (C1Qtnf1/Ctrp1) Elisa Kit
Supplier:  Biomatik Corporation EKC33245
Encompass_Preferred
Human Complement C1Q Tumor Necrosis Factor-Related Protein 1 (C1Qtnf1/Ctrp1) Elisa Kit for detecting C1Q And Tumor Necrosis Factor Related Protein 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9BXJ1; C1QTNF1. Target Synonyms: C1q and tumor necrosis factor related protein 1; C1QT1_HUMAN; C1QTNF1; Complement C1q tumor necrosis factor related protein 1; Complement C1q tumor necrosis factor-related protein 1; CTRP 1; CTRP1; FLJ90694; G protein coupled receptor interacting protein 1; G protein-coupled receptor-interacting protein; GIP; ZSIG37. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3885