Learn More
Biomatik Corporation Human Complement Component C6 (C6) Elisa Kit
Supplier:  Biomatik Corporation EKC33261
Encompass_Preferred
Human Complement Component C6 (C6) Elisa Kit for detecting Complement Component 6 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P13671; C6. Target Synonyms: AW111623; C6; CO6_HUMAN; Complement component 6; Complement component C6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3889