Learn More
Biomatik Corporation Human Complement Factor H-Related Protein 2 (Cfhr2) Elisa Kit
Supplier:  Biomatik Corporation EKC33267
Encompass_Preferred
Human Complement Factor H-Related Protein 2 (Cfhr2) Elisa Kit for detecting Complement Factor H-Related 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 1.56ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P36980; CFHR2. Target Synonyms: CFHL2; CFHR2; Complement factor H related protein 2; Complement factor H-related 2; Complement factor H-related protein 2; DDESK59; Factor H-related gene 2; FHR 2; FHR-2; FHR2; FHR2_HUMAN; H factor (complement)-like 3; H factor like 3; H factor like protein 2; H factor-like 3; H factor-like protein 2; HFL3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3893