Learn More
Biomatik Corporation Human Cx3C-Chemokine Receptor 1, Cx3Cr1 Elisa Kit
Supplier:  Biomatik Corporation EKC33332
Encompass_Preferred
Human Cx3C-Chemokine Receptor 1, Cx3Cr1 Elisa Kit for detecting Chemokine (C-X3-C Motif) Receptor 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P49238; CX3CR1. Target Synonyms: CX3CR1; CMKBRL1; GPR13; CX3C chemokine receptor 1; C-X3-C CKR-1; CX3CR1; Beta chemokine receptor-like 1; CMK-BRL-1; CMK-BRL1; Fractalkine receptor; G-protein coupled receptor 13; V28. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3899