Learn More
Biomatik Corporation Human Cytochrome-C, Cyt-C Elisa Kit
Supplier:  Biomatik Corporation EKC33386
Encompass_Preferred
Human Cytochrome-C, Cyt-C Elisa Kit for detecting Cytochrome C, Somatic in serum, plasma, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500ng/ml. Sensitivity: 1.95ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P99999; CYCS. Target Synonyms: CYC; CYC_HUMAN; CYCS; Cytochrome c; Cytochrome c somatic; HCS; THC4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3910