Learn More
Biomatik Corporation Human Cytoskeleton-Associated Protein 4 (Ckap4) Elisa Kit
Supplier:  Biomatik Corporation EKC33402
Encompass_Preferred
Human Cytoskeleton-Associated Protein 4 (Ckap4) Elisa Kit for detecting Cytoskeleton-Associated Protein 4 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.44-1500pg/ml. Sensitivity: 5.86pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q07065; CKAP4. Target Synonyms: 63 kDa membrane protein; CKAP 4; Ckap4; CKAP4_HUMAN; Cytoskeleton associated protein 4; Cytoskeleton-associated protein 4; ERGIC 63; ERGIC63; MGC99554; p63; Transmembrane protein 63kD endoplasmic reticulum Golgi intermediate compartment; Type II transmembrane protein p63. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3914