Learn More
Biomatik Corporation Human Dehydroepiandrosterone Sulfate, Dhea-S Elisa Kit
Supplier:  Biomatik Corporation EKC33423
Encompass_Preferred
Human Dehydroepiandrosterone Sulfate, Dhea-S Elisa Kit for detecting Dehydroepiandrosterone Sulfate, Dhea-S in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 20-4000ng/ml. Sensitivity: 10ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3918