Learn More
Biomatik Corporation Human Delta-Like Protein 3 (Dll3) Elisa Kit
Supplier:  Biomatik Corporation EKC33429
Encompass_Preferred
Human Delta-Like Protein 3 (Dll3) Elisa Kit for detecting Delta-Like 3 (Drosophila) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 18.75-1200pg/ml. Sensitivity: 4.68pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NYJ7; DLL3. Target Synonyms: Delta Drosophila like 3; Delta like 3 Drosophila; Delta like 3 homolog Drosophila; Delta like 3 protein; Delta like protein 3 precursor; Delta-like protein 3; Delta3; Dll3; DLL3_HUMAN; Drosophila Delta homolog 3; SCDO1; SCOD1; Spondylocostal dysostosis autosomal recessive. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3920