Learn More
Biomatik Corporation Human Deoxyribonuclease Gamma (Dnase1L3) Elisa Kit
Supplier:  Biomatik Corporation EKC33435
Encompass_Preferred
Human Deoxyribonuclease Gamma (Dnase1L3) Elisa Kit for detecting Deoxyribonuclease I-Like 3 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.44-1500pg/ml. Sensitivity: 5.86pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13609; DNASE1L3. Target Synonyms: Deoxyribonuclease gamma; Deoxyribonuclease I like 3; Deoxyribonuclease I like III; Deoxyribonuclease I-like 3; DHP 2; DHP2; DNAS1L3; DNase gamma; DNase I homolog protein 2; DNase I homolog protein DHP2; DNase I like 3; DNase I-like 3; DNASE1L3; DNSL3_HUMAN; Liver and spleen DNase; LS DNase; LS-DNase; LSD; SLEB 16; SLEB16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3923