Learn More
Biomatik Corporation Human Diamine Acetyltransferase 1 (Sat1) Elisa Kit
Supplier:  Biomatik Corporation EKC33451
Encompass_Preferred
Human Diamine Acetyltransferase 1 (Sat1) Elisa Kit for detecting Spermidine/Spermine N1-Acetyltransferase 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P21673; SAT1. Target Synonyms: DC21; Diamine acetyltransferase 1; Diamine N acetyltransferase 1; EC 2.3.1.57; KFSD; KFSDX; Polyamine N acetyltransferase 1; Polyamine N-acetyltransferase 1; Putrescine acetyltransferase; SAT; SAT1; SAT1_HUMAN; Spermidine/spermine N (1) acetyltransferase 1; Spermidine/spermine N (1)-acetyltransferase 1; Spermidine/spermine N1 acetyltransferase 1; Spermidine/spermine N1 acetyltransferase alpha; SSAT 1; SSAT; SSAT-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3927