Learn More
Biomatik Corporation Human Dipeptidase 1 (Dpep1) Elisa Kit
Supplier:  Biomatik Corporation EKC33463
Encompass_Preferred
Human Dipeptidase 1 (Dpep1) Elisa Kit for detecting Dipeptidase 1 (Renal) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.86pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P16444; DPEP1. Target Synonyms: Dehydropeptidase-I; Dipeptidase 1 (renal); Dipeptidase 1; DPEP1; DPEP1_HUMAN; hRDP; MBD1; MDP; Microsomal dipeptidase; RDP; Renal dipeptidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3929