Learn More
Biomatik Corporation Human Dna- (Apurinic Or Apyrimidinic Site) Lyase (Apex1) Elisa Kit
Supplier:  Biomatik Corporation EKC33490
Encompass_Preferred
Human Dna- (Apurinic Or Apyrimidinic Site) Lyase (Apex1) Elisa Kit for detecting Apex Nuclease (Multifunctional Dna Repair Enzyme) 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P27695; APEX1. Target Synonyms: AP endonuclease 1; AP endonuclease class I; AP lyase; APE 1; APE; APE-1; APEN; APEX 1; APEX; APEX nuclease (multifunctional DNA repair enzyme) 1; Apex nuclease 1; APEX nuclease; APEX1; APEX1_HUMAN; Apurinic endonuclease; Apurinic-apyrimidinic endonuclease 1; Apurinic/apyrimidinic (abasic) endonuclease; Apurinic/apyrimidinic endonuclease 1; Apurinic/apyrimidinic exonuclease; APX; BAP1; Deoxyribonuclease (apurinic or apyrimidinic); DNA (apurinic or apyrimidinic site) lyase; DNA-(apurinic or apyrimidinic site) lyase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3931