Learn More
Biomatik Corporation Human E3 Ubiquitin-Protein Ligase Trim63 (Trim63) Elisa Kit
Supplier:  Biomatik Corporation EKC33524
Encompass_Preferred
Human E3 Ubiquitin-Protein Ligase Trim63 (Trim63) Elisa Kit for detecting Tripartite Motif-Containing 63 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-800pg/ml. Sensitivity: 3.1pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q969Q1; TRIM63. Target Synonyms: E3 ubiquitin-protein ligase TRIM63; FLJ32380; IRF; Iris RING finger protein; MURF 1; MURF-1; MuRF1; MURF2; Muscle specific ring finger protein 1; Muscle specific ring finger protein 2; Muscle-specific RING finger protein 1; OTTHUMP00000008701; RING finger protein 28; RNF 28; RNF28; SMRZ; Striated muscle RING zinc finger protein; TRI63_HUMAN; TRIM 63; Tripartite motif containing 63; tripartite motif containing 63; E3 ubiquitin protein ligase; Tripartite motif containing protein 63; Tripartite motif-containing protein 63; Ubiquitin ligase TRIM63. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3935