Learn More
Biomatik Corporation Human Ecto-Nox Disulfide-Thiol Exchanger 2 (Enox2) Elisa Kit
Supplier:  Biomatik Corporation EKC33532
Encompass_Preferred
Human Ecto-Nox Disulfide-Thiol Exchanger 2 (Enox2) Elisa Kit for detecting Ecto-Nox Disulfide-Thiol Exchanger 2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q16206; ENOX2. Target Synonyms: APK1; APK1 antigen; COVA1; Cytosolic ovarian carcinoma antigen 1; Ecto NOX disulfide thiol exchanger 2; ENOX 2; ENOX2; ENOX2_HUMAN; OTTHUMP00000024021; OTTHUMP00000024022; Protein disulfide-thiol oxidoreductase; tNOX; Tumor associated hydroquinone oxidase; Tumor-associated hydroquinone oxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3937