Learn More
Biomatik Corporation Human Endostatin, Es Elisa Kit
Catalog No. 502293943
Encompass_Preferred
Human Endostatin, Es Elisa Kit for detecting Endostatin, Es in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: FLJ27325; FLJ34914; KNO; KNO1; MGC74745; alpha 1 type XVIII collagen; antiangiogenic agent; endostatin; multi-functional protein MFP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-3943 Supplier Biomatik Corporation Supplier No. EKC33564
Human Endostatin, Es Elisa Kit for detecting Endostatin, Es in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: FLJ27325; FLJ34914; KNO; KNO1; MGC74745; alpha 1 type XVIII collagen; antiangiogenic agent; endostatin; multi-functional protein MFP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.