Learn More
Biomatik Corporation Human Enteropeptidase (Prss7) Elisa Kit
Supplier:  Biomatik Corporation EKC33576
Encompass_Preferred
Human Enteropeptidase (Prss7) Elisa Kit for detecting Protease, Serine, 7 (Enterokinase) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P98073; TMPRSS15. Target Synonyms: TMPRSS15; ENTK; PRSS7; Enteropeptidase; Enterokinase; Serine protease 7; Transmembrane protease serine 15. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3946